- Protein/Peptide Type:Recombinant
- Species:Mouse
- Size:0.05 mg
- Tag sequence:C-6 His tag
- Storage Conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Endotoxin Content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Protein Synonyms:GDF-8|Mstn|Growth/differentiation factor 8|Myostatin|Gdf8
- Protein/Peptide Name:GDF8
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular Weight:28.4 kD
- Sequence:Asn25-Ser265
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested Applications:Bioassay
- Cat. No.:75789-910
- Supplier no.:91-625
Mouse Growth/differentiation factor 8(Mstn, GDF-8) is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. It is expressed specifically in developing and adult skeletal muscle. It exists as a homodimer, and interacts with WFIKKN2, leading to inhibit its activity. This protein can act specifically as a negative regulator of skeletal muscle growth. It regulates cell growth and differentiation in both embryonic and adult tissues. Experiments in mice have improved that GDF-8 is a key regulator of mesenchymal stem cell proliferation and differentiation, and mice lacking Myostatin encoding gene show decreased body fat and a generalized increase in bone density and strength.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.