Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant CCL24 (from E. coli)
Mouse Recombinant CCL24 (from E. coli)
Catalog # 75789-838
Supplier:  Prosci
CAS Number:  
Mouse Recombinant CCL24 (from E. coli)
Catalog # 75789-838
Supplier:  Prosci
Supplier Number:  91-581
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    0.05 mg
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein Synonyms:
    C-C motif chemokine 24|Small-inducible cytokine A24|Ccl24|Eotaxin-2|Eosinophil chemotactic protein 2|Scya24
  • Protein/Peptide Name:
    CCL24
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    10.3 kD
  • Sequence:
    Val27-Val119
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75789-838
  • Supplier no.:
    91-581

Specifications

About this item

Mouse CCL24 is a secreted protein, which is a member of the CC chemokine subfamily. Mouse Ccl24 cDNA encodes a 119 amino acid residue precursor protein, shares approximately 58% amino acid sequence identity with human Ccl24. It is predominantly expressed in the jejunum and spleen and also be induced in the lung by allergen challengeand IL4. Mouse ccl24 has lower chemotactic activity for neutrophils but none for monocytes and activated lymphocytes. Ccl24 is chemotactic for resting T-lymphocytes, eosinophils and can bind to CCR3. LPS and IL4 also differentially regulate the expression of Ccl24 in monocytes and macrophages.

This recombinant protein can be used for biological assays. For research use only.