Order Entry
United States
Orders LinkContactUsLinkComponent
Human Recombinant MOB1A (from <i>E. coli</i>)

Human Recombinant MOB1A (from E. coli)

0.05 mg
Catalog # 75789-230
Supplier:   Prosci
Supplier #:  91-277
Human Recombinant MOB1A (from <i>E. coli</i>)
Catalog # 75789-230
Supplier:  Prosci
CAS Number:  
Human Recombinant MOB1A (from E. coli)

0.05 mg

CAS Number:  
Price
$958.78
Log in for availability
Each (50µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

MOB Kinase Activator 1A (MOB1A) belongs to the MOB1/phocein family, which acts as an activator of LATS1/2 in the Hippo signaling pathway, plays a key role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. MOBKL1B stimulates the kinase activity of STK38 and STK38L. MOBKL1B binds to and regulate downstream targets such as the NDR-family protein kinases and LATS1 kinase.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Documents

Specifications

Specifications

  • Size:
    0.05 mg
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Molecular Weight:
    26 kD
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Tested Applications:
    Bioassay
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Sequence:
    Ser2-Arg216
  • Source:
    E. coli
  • Cat. No.:
    75789-230
  • Protein/Peptide Type:
    Recombinant
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM Tris, 0.15M NaCl, pH 8.0 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Protein Synonyms:
    MOB4B|Mob1 Homolog 1B|C2orf6|MOB Kinase Activator 1A|Mob1 Alpha|MOBKL1B|Mps One Binder Kinase Activator-like 1B|Mob1A|MOBK1B
  • Protein/Peptide Name:
    MOB1A
  • Tag sequence:
    C-6 His tag
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.