Order Entry
United States
Orders LinkContactUsLinkComponent
Human Recombinant HSF2 (from <i>E. coli</i>)

Human Recombinant HSF2 (from E. coli)

0.05 mg
Catalog # 75789-204
Supplier:   Prosci
Supplier #:  91-264
Human Recombinant HSF2 (from <i>E. coli</i>)
Catalog # 75789-204
Supplier:  Prosci
CAS Number:  
Human Recombinant HSF2 (from E. coli)

0.05 mg

CAS Number:  
Price
$958.78
Log in for availability
Each (50µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Heat Shock Factor Protein 2 (HSF2) belongs to the HSF family of transcription factors that bind specifically to the heat-shock promoter element and activate transcription. In higher eukaryotes, HSF is unable to bind to the HSE unless the cells are heat shocked. HSF2 is widely expressed in many cells and tissues. HSF2 is located on Cytoplasmic during normal growth. But when it is activited, HSF2 moves to the nucleus.

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Documents

Specifications

Specifications

  • Size:
    0.05 mg
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Molecular Weight:
    15.9 kD
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Tested Applications:
    Bioassay
  • Endotoxin Content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Sequence:
    Ser411-Ser536
  • Source:
    E. coli
  • Cat. No.:
    75789-204
  • Protein/Peptide Type:
    Recombinant
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM DTT, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Protein Synonyms:
    HSF2|HSTF2|Heat Shock Transcription Factor 2|Heat Shock Factor Protein 2|HSTF 2|HSF 2
  • Protein/Peptide Name:
    HSF2
  • Tag sequence:
    N-6 His tag
  • Species:
    Human

Specifications