Order Entry
United States
ContactUsLinkComponent
Human Recombinant LR3-IGF-1 (from E. coli)
Human Recombinant LR3-IGF-1 (from E. coli)
Catalog # 75788-758
Supplier:  Prosci
CAS Number:  
Human Recombinant LR3-IGF-1 (from E. coli)
Catalog # 75788-758
Supplier:  Prosci
Supplier Number:  91-017
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage Conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Endotoxin Content:
    <0.05 ng/ug (0.5 IEU/ug) as determined by LAL test.
  • Biological Activity:
    ED50 is greater than 200 ng/ml as determined by an IGF binding protein assay.
  • Protein Synonyms:
    IBP1|IGF1|Mechano Growth Factor|IGF-I|Insulin-like Growth Factor I|MGF|Somatomedin-C
  • Protein/Peptide Name:
    LR3-IGF-1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular Weight:
    9.1 kD
  • Sequence:
    Gly49-Ala118
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 300mM HAc-NaAc, pH 6.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in 500mM Acetic Acid. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested Applications:
    Bioassay
  • Cat. No.:
    75788-758
  • Supplier no.:
    91-017

Specifications

About this item

Insulin-like growth factor I (IGF1) belongs to the family of insulin-like growth factors that are structurally homologous to proinsulin. Mature IGFs are generated by proteolytic processing of inactive precursor proteins, which contains the N- and C-terminal propeptide regions. Mature human IGF-I consisting of 70 amino acids has 94% identity with mouse IGF-I and exhibits cross-species activity. IGF-1 binds IGF-IR, IGF-IIR, and the insulin receptor and plays a key role in cell cycle progression, cell proliferation and tumor progression. IGF-1 expression is regulated by growth hormone. R3 IGF-1 is an 83 amino acid analog of IGF-1 comprising the complete human IGF-1 sequence with the substitution of an Arg (R) for the Glu(E) at position three, hence R3, and a 13 amino acid extension peptide at the N terminus. R3 IGF-1 has been produced with the purpose of increasing biological activity. R3 IGF-1 is significantly more potent than human IGF-I in vitro.

This recombinant protein can be used for biological assays. For research use only.