Specifications
- Antibody Type:Primary
- Antigen Name:vasodilator-stimulated phosphoprotein
- Antigen Symbol:VASP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:P50552
- Antigen Synonyms:VASP
- Storage Buffer:Liquid. Crude serum containing 0.02% sodium azide.
- Storage Temperature:Stable for at least 1 year after receipt when stored at –20 °C.
- Shipping Temperature:4 °C
- Immunogen:Recombinant human VASP fused to a His-tag.
- Tested Applications:Immunocytochemistry
- Cat. No.:75793-494
- Supplier no.:36-349
Specifications
About this item
VASP (vasodilator-stimulated phosphoprotein) is a proline-rich protein substrate of cAMP- and cGMP-dependent protein kinases. VASP is an actin-associated protein involved in a range of processes dependent on cytoskeleton remodeling and cell polarity such as axon guidance, lamellipodial and filopodial dynamics, platelet activation and cell migration. VASP promotes actin filament elongation. It protects the barbed end of growing actin filaments against capping and increases the rate of actin polymerization in the presence of capping proteins. VASP stimulates actin filament elongation by promoting the transfer of profilin-bound actin monomers onto the barbed end of growing actin filaments and it plays a role in actin-based mobility of Listeria monocytogenes in host cells. It regulates actin dynamics in platelets and plays an important role in regulating platelet aggregation. VASP phosphorylation is used to monitor the effect of so-called antiplatelet drugs that reduce platelet reactivity and are used to prevent stent thrombosis, strokes and heart attacks in patients. Phosphorylation of VASP at Ser157 causes a mobility shift in SDS gel electrophoresis from 46 to 50kDa, which has been used as a convenient marker to monitor cyclic nucleotide-dependent protein kinase activity.
ELISA: (direct or indirect: 1:2,000-1:5,000). Western blot: (1:2,000-1:5,000 using ECL. Suggested blocking and dilution buffer is PBST with 0.05% Tween 20 and 5% skim milk. Suggested incubation time is 1 hour at rooom temperature). Optimal conditions should be determined individually for each application.
Type: Primary
Antigen: VASP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human