Revco® PLUS CxF Series Ultra-Low Temperature Chest Freezers, -40°C, Thermo Scientific
Supplier: Avantor
Certificates
About this item
Reliably store samples with versatile ultra-low temperature chest freezers.
- Eye-level, easy-to-use microprocessor control panel
- Stainless steel interior
- Easy-to-remove air filter
- Access code to ensure setpoint security
- 5″ (127mm) foamed-in-place, polyurethane insulation
- Ergonomic, lockable door handle
- Optional built-in chart recorder or data logger for continuous monitoring
- Message center: 12-character alphanumeric LED displays setpoints, system status and alarm messages
- Heavy gauge, cold-rolled steel exterior with a powder coat paint finish that resists chipping and rust
Convenient interior covers minimize sample exposure to outside conditions. Inner sub-lids protect bulk samples from ambient exposure during single sample retrievals. Optional CO2 or LN2 safety back-up system is available for additional protection in the event of a power or mechanical failure. RS-232 and 4-20 milliamp analog outputs are included for independent monitoring devices and recorders. Freezer includes heavy-duty casters with locking tandem wheel design for smooth mobility.
Frequently Bought Together