- Description:PSA (Prostate Specific Antigen) Reagent
- Size:96 Tests
- Cat. No.:10023-508
- Format:Liquid
- Storage temperature:2...8°C (35.6...46.4 °F)
- Supplier no.:TM1019-96
PSA (Prostate Specific Antigen) Reagent Set is used for the quantitative determination of the Prostate Specific Antigen (PSA) in serum.
- Quantitative determination of the Prostate Specific Antigen (PSA) in serum
- ELISA test is based on the principle of a solid phase enzyme-linked immunosorbent assay
- Concentration of PSA is directly proportional to the color intensity of the test sample
PSA (Prostate Specific Antigen) Reagent Set is used for the quantitative determination of the Prostate Specific Antigen (PSA) in serum
A solution of TMB Reagent is added and incubated at room temperature for 20 minutes, resulting in the developing of a blue color. The color development is stopped with the addition of Stop Solution changing the color to yellow. The concentration of PSA is directly proportional to the color intensity of the test sample. Absorbance is measured spectrophotometrically at 450nm.
For research use only in the USA. The PSA ELISA test is based on the principle of a solid phase enzyme-linked immunosorbent assay. The assay system utilizes a goat anti-PSA antibody directed against intact PSA for solid phase immobilization (on the microtiter wells). A monoclonal anti-PSA antibody conjugated to horseradish peroxidase (HRP) is in the antibody-enzyme conjugate solution. The test sample is allowed to react first with the immobilized goat antibody at room temperature for 60 minutes. The wells are washed to remove any unbound antigen. The monoclonal anti-PSA-HRP conjugate is then reacted with the immobilized antigen for 60 minutes at room temperature resulting in the PSA molecules being sandwiched between the solid phase and enzyme-linked antibodies. The wells are washed with water to remove unbound-labeled antibodies.