Triglycerides - Liquid Reagent Set, Pointe Scientific
Supplier: HORIBA INSTRUMENTS INCORPORATED
Certificates
About this item
Triglycerides - Liquid Reagent Set is used for the in vitro quantitative determination of Triglycerides in serum or plasma.
- In vitro quantitative determination of Triglycerides in serum or plasma
- Intensity of the colored complex formed is directly proportional to the triglycerides concentration
- Diagnostic use only
The Hitachi 780 mL reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are (12) 65 mL Reagent bottles in this kit.
The Hitachi 1 L reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are (10) 100 mL Reagent bottles in this kit.
The Olympus reagent is for use on the Beckman Coulter AU 400, 480, and 640 analyzers and is for in vitro diagnostic use only. There are (12) 65 mL Reagent bottles in this kit.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
An NIST-traceable Triglycerides standard or serum calibrator and two levels of Commercially available controls.
An NIST-traceable Triglycerides standard or serum calibrator and two levels of Commercially available controls.
Frequently Bought Together