Order Entry
United States
ContactUsLinkComponent
Triglycerides Standard, Pointe Scietnific
Triglycerides Standard, Pointe Scietnific
Catalog # 10022-966
CAS Number:  
Triglycerides Standard, Pointe Scietnific
Catalog # 10022-966
Supplier Number:  T7531-STD
CAS Number:  

Specifications

  • Concentration:
    200 mg/dl
  • Description:
    Triglycerides Standard
  • Size:
    15 mL
  • Cat. No.:
    10022-966
  • Format:
    Liquid
  • Storage temperature:
    2...8°C (35.6...46.4 °F)
  • Supplier no.:
    T7531-STD

Specifications

About this item

Triglycerides Standard is Intended for use as a triglycerides standard for standardization or calibration of enzymatic triglycerides procedures.

  • Standardization or calibration of enzymatic triglycerides procedures
  • Should be used to calibrate the determination of unknown specimens
  • Enzymatic triglycerides assay used

Triglycerides Standard is Intended for use as a triglycerides standard for standardization or calibration of enzymatic triglycerides procedures. The standard should be used to calibrate the determination of unknown specimens using an enzymatic triglycerides assay. Controls should be included to validate the calibration of the assay.

Certifications: CE Certified
Ordering information: Materials Required but not Provided:
Triglycerides Standards are intended to be used with Pointe Scientific Triglycerides Reagents.