Uric Acid (Hitachi & Beckman) Reagent Set, Pointe Scientific
Supplier: HORIBA INSTRUMENTS INCORPORATED
Certificates
About this item
Uric Acid (Hitachi & Beckman) Reagent Set is for the quantitative determination of Uric Acid in serum.
- Quantitative determination of Uric Acid in serum
Uric Acid (Hitachi & Beckman) Reagent Set is for the quantitative determination of Uric Acid in serum
The Hitachi 729 mL reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are nine 65 mL R1 Reagent bottles and nine 16 mL R2 Reagent bottles in this kit.
The Hitachi 615 mL reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are five 100 mL R1 Reagent bottles and five 23 mL R2 Reagent bottles in this kit.
Certifications: CE Certified
Ordering information: Materials Required but not Provided:
A standard or serum calibrator. Serum controls with known normal and abnormal uric acid values.
A standard or serum calibrator. Serum controls with known normal and abnormal uric acid values.
Frequently Bought Together