About this item
The Gamma Glutamyl Transferase (Hitachi & Beckman) Reagent Set is for the quantitative kinetic determination of gamma glutamyl transferase (GGT) activity in serum.
- Quantitative kinetic determination of gamma glutamyl transferase (GGT) activity in serum
- Amount of 5-amino-2-nitrobenzoate formed is proportional to γ-GT activity and may be measured kinetically
The Gamma Glutamyl Transferase (Hitachi & Beckman) Reagent Set is for the quantitative kinetic determination of gamma glutamyl transferase (GGT) activity in serum
The Hitachi 295 mL reagent is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are five 42 mL R1 Reagent bottles and five 17 mL R2 Reagent bottles in this kit.
The Hitachi 315 mL reagent is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are five 45 mL R1 Reagent bottles and two 45 mL R2 Reagent bottles in this kit.
Serums with known normal and abnormal GGT values.