About this item
The Calcium Reagent Set is for the quantitative determination of Calcium in serum.
- Quantitative determination of Calcium in serum
The Calcium Reagent Set is for the quantitative determination of Calcium in serum
The Hitachi 576 mL reagent is for use on the Hitachi 917 analyzer only. There are six 66 mL R1 Reagent bottles and six 30 mL R2 Reagent bottles in this kit.
The Hitachi 1L reagent is for use on Hitachi 717, 911, 912, and 914 analyzers. There are seven 100 mL R1 Reagent bottles and three 100 mL R2 Reagent bottles in this kit.
Control sera with known normal and abnormal calcium values as well as Calibrator or Standard.