Dualmat™ Static Dissipative Mats, Precut, Royal Blue, ACL
Supplier: ACL Staticide
Certificates
About this item
Dualmat™ is a heavy duty ESD-safe rubber material designed for use on tables and grounded work bench surfaces
These dual layer precut mats feature two 10mm female snaps at the top and bottom left side. Snaps can be located elsewhere if specified. They are available in 8 standard sizes, and custom sizes are also available. Dualmats™ are low outgassing and low VOC, making them ideal for use in cleanrooms and other controlled environments. Additionally, they are UV protected so the color will not fade.
Dissipative on the colored side and conductive on the back side, the design makes it ideal for use with constant monitors. Embossed finish reduces glare, Resistance is 10⁶ to 10⁸ ohms. Electricals warranted for the lifetime of the mat.
Certifications: Compliant to ANSI/ESD S20.20 and RoHS.
Frequently Bought Together