

2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Avantor
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89.5 °C
Density:0.785 g/cm³
Flash Pt:12 °C
Storage Temperature:Ambient
UN:1219
ADR:3,II
:GHS07
:GHS02
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA safer chemical solvent (Green circle rating); Chem21 solvent selection guide recommended by default and after discussion
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.2 % (K.F.)
- Boiling point:81 to 83 °C
- Density:(20/20 °C) 0.784 to 0.787
- Solubility:(in water) Passes test
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Carbonyl compounds:≤0.002 % (propionaldehyde)
- Titrable acid or base:≤0.0001 meq/g
Specification Test Results
Frequently Bought Together