Order Entry
United States
Orders LinkContactUsLinkComponent
3-(4-Bromophenyl)propionic acid 97%

3-(4-Bromophenyl)propionic acid 97%

Irritant
Marketsource
3-(4-Bromophenyl)propionic acid 97%
Supplier:  
Marketsource
Certificates

0 Options Available Below

Product Details & Documents
  • CAS Number:1643-30-7
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD01310793
  • Molecular Weight:229.07
  • Melting Pt:133…136 °C
  • Storage Temperature:Ambient
  • UN:3335
  • ADR:9,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to yellow to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    134 to 138 °C
  • HPLC:
    ≥96.0 %
Product Family Options
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.