Order Entry
United States
ContactUsLinkComponent
 
Zinc Standard Solution, for AAS, 1 mg/ml Zn in 2% HCl, Thermo Scientific
Catalog # 200058-858
Supplier:  Avantor
CAS Number:  
Zinc Standard Solution, for AAS, 1 mg/ml Zn in 2% HCl, Thermo Scientific
Catalog # 200058-858
Supplier:  Avantor
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    2% HCl
  • Pack type:
    Plastic bottle
  • Size:
    500 ml
  • Cat. No.:
    200058-858
  • Element:
    Zinc
  • Supplier no.:
    AC19646-5000|19646-5000

Specifications

About this item

CAS No.:

MDL No.: MFCD00011291
UN: 3264
Hazard Class: 8
Packing group: III
GHS Symbols: GHS05
Density: 1.01
Boiling Point: 100°C -
Appearance: Clear colorless Solution
Storage Temperature: Ambient
Signal word: Warning