Alkaline Phosphatase Reagent Set, Pointe Scientific
Supplier: HORIBA INSTRUMENTS INCORPORATED
Certificates
About this item
The Alkaline Phosphatase Reagent Set is used for the quantitative determination of alkaline phosphatase in human serum.
- Quantitative determination of alkalin phosphatase in human serum
- In vitro diagnostic use only
For the quantitative determination of alkaline phosphatase in human serum
The Hitachi 302 mL kit is for use on the Hitachi 917 analyzer only and for in vitro diagnostic use only. There are five 50 mL R1 Reagent bottles and four 13 mL R2 Reagent bottles in this kit.
The Hitachi 492 mL kit is for use on Hitachi 717, 911, 912, and 914 analyzers and for in vitro diagnostic use only. There are four 100 mL R1 Reagent bottles and four 23 mL R2 Reagent bottles in this kit.
Certifications: CE Compliant
Ordering information: Materials Required but not Provided:
The validity of the reaction should be monitored using control sera with known normal and abnormal ALP activities (C7590/91/92)
The validity of the reaction should be monitored using control sera with known normal and abnormal ALP activities (C7590/91/92)
Frequently Bought Together