Ac-DEVD-CHO Caspase-3 Inhibitor
Supplier: Biotium
Certificates
About this item
Ac-DEVD-CHO can be used to inhibit Caspase-3/7 activity in apoptotic cells.
- Inhibit Caspase-3/7 activity in apoptotic cells
- Study events downstream of Caspase-3/7 activation
- Synthetic tetrapeptide competitive inhibitor for Caspase-3/7
This can be used to inhibit Caspase-3/7 activity in apoptotic cells, and to study events downstream of Caspase-3/7 activation. Ac-DEVD-CHO is a synthetic tetrapeptide competitive inhibitor for Caspase-3/7, and contains the amino acid sequence of the PARP cleavage site.
Specifications
- Protein/Peptide Name:Ac-DEVD-CHO Caspase-3 Inhibitor