To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:5 KU
- Enzyme Name:Salt Activated Nuclease
- Cat. No.:10147-170
- Supplier no.:1B1664-5KU
Specifications
About this item
VWR® Salt Active Nuclease (SAN) is a very active non-specific endonuclease from Pichia pastoris that cleaves double- and single-stranded DNA and RNA
- Store frozen (-20 – 0°C)
- Stable 6 months stored cold (2 – 8°C) and two years frozen (-20 – 0°C)
- SAN tolerates multiple freeze-thaws
It is active at above neutral pH (optimal pH 8.5) over a wide temperature range (10- 40°C), and unlike other nucleases it has optimum activity at high concentrations of salt (0.5M NaCl). SAN is ideal for use in removal of DNA and RNA from cell extracts and protein samples. Thus, it is very useful in recombinant protein purifications.