$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Organic Standards
Creatinine Standard Set
Creatinine Standard Set
10022-646
:
Avantor
:
Creatinine Standard Set
10022-646
:
Avantor
:
Formula:
C₄H₇N₃O
Description:
Creatinine Standard
Storage temperature:
15-30 °C