Order Entry
United States
ContactUsLinkComponent
 
Streptavidin, Alexa Fluor® 647, Jackson ImmunoResearch
Catalog # 102643-996
CAS Number:  
Streptavidin, Alexa Fluor® 647, Jackson ImmunoResearch
Catalog # 102643-996
Supplier Number:  016-600-084
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle, amber
  • Conjugation:
    Alexa Fluor® 647
  • Source:
  • Species:
  • Size:
    1 mg
  • Environmentally Preferable:
  • Storage Conditions:
    4 °C
  • Protein/Peptide Name:
    Streptavidin
  • Concentration:
    1.4 - 1.8 mg/mL
  • Formulation:
    Freeze-dried powder, 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 15mg/mL BSA, 0.05% Azide
  • Shipping Temperature:
    ambient
  • Cat. No.:
    102643-996
  • Supplier no.:
    016-600-084

Specifications

About this item

Steptavidin, a bacterial protein isolated from Streptomyces avidinii, is similar to egg-white avidin in its ability to bind biotin, and has been used as a replacement for egg-white avidin because of its more favorable chemical properties.
Conjugates of streptavidin are recommended for use with Biotin-SP-conjugated antibodies and Biotin-SP-conjugated ChromPure proteins.