Order Entry
United States
Orders LinkContactUsLinkComponent
Rat Transferrin

Rat Transferrin

Glass bottle, amber, 5 mg
Catalog # 102643-816
Marketsource
Supplier #:  012-000-050
Rat Transferrin
Catalog # 102643-816
Marketsource
CAS Number:  
Rat Transferrin

Glass bottle, amber, 5 mg

CAS Number:  
Price
$193.23
Log in for availability
Each (5mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

ChromPure is a trade name for highly purified proteins from the serum of non-immunized animals.

Specifications

Specifications

  • Pack type:
    Glass bottle, amber
  • Size:
    5 mg
  • Storage Conditions:
    4 °C
  • Cat. No.:
    102643-816
  • Molecular Weight:
    ~ 80 kDa
  • Purification:
    Chromatographically Purified
  • Shipping Temperature:
    ambient
  • Formulation:
    Sterile-filtered liquid, 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6
  • Protein/Peptide Name:
    Transferrin
  • Conjugation:
    Unconjugated
  • Concentration:
    5.0 - 6.0 mg/mL
  • Species:
    Rat

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.