Order Entry
United States
Orders LinkContactUsLinkComponent
Human Transferrin

Human Transferrin

Glass bottle, amber, 5 mg
Catalog # 102643-694
Marketsource
Supplier #:  009-000-050
Human Transferrin
Catalog # 102643-694
Marketsource
CAS Number:  
Human Transferrin

Glass bottle, amber, 5 mg

CAS Number:  
Price
$193.23
Log in for availability
Each (5mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

ChromPure is a trade name for highly purified proteins from the serum of non-immunized animals.

Specifications

Specifications

  • Pack type:
    Glass bottle, amber
  • Size:
    5 mg
  • Storage Conditions:
    4 °C
  • Cat. No.:
    102643-694
  • Molecular Weight:
    ~ 80 kDa
  • Purification:
    Chromatographically Purified
  • Shipping Temperature:
    ambient
  • Formulation:
    Sterile-filtered liquid, 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6
  • Protein/Peptide Name:
    Transferrin
  • Conjugation:
    Unconjugated
  • Concentration:
    5.0 - 6.0 mg/mL
  • Species:
    Human

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.