PIG® Surgical Floor Skimmer
Supplier: New Pig
Certificates
About this item
Revolutionary design attacks puddle problems from all sides to help keep O.R. floors dry and safe.
- Cloverleaf shape directs liquids into the suction channel for maximum efficiency and performance
- Proprietary bottom wicking pad removes 60 liters per hour
- Top fluid collection channel captures liquid from the drapes and channels it to the wicking pad
- Composition: Polypropylene/Cellulose Blend
- Thin Low-profile design
Easy-slide floor pad can be repositioned or swept across the floor with your foot and captures liquids no matter where they go. Silent suction design helps keep the operating room quite. Suction tubing (included) allows access to all areas around the O.R. table.
Patent-pending design.
Accessories information: Latex-free suction tubing 3.66 mm.
Ordering information: Sold as 10 or 20 per box.
Frequently Bought Together