≥92% pure active human Recombinant TNFR1 / CD12a / TNFRSF1A
Tumor necrosis factor receptor 1 (TNF-R1) also known as Tumor necrosis factor receptor superfamily member 1A (TNFRSF1A), TNFAR, CD antigen CD12a, belongs to the tumor necrosis factor receptor superfamily. TNF-R1 contains one death domain and four TNFR-Cys repeats. TNF-R1 is the receptor of TNFSF2 / TNF-alpha and homotrimeric TNFSF1 / lymphotoxin - alpha. The adapter molecule FADD recruits caspase-8 to the activated receptor. The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. TNF-R1 contributes to the induction of non - cytocidal TNF effects including anti-viral state and activation of the acid sphingomyelinase. Defects in TNFRSF1A are the cause of familial Hibernian fever (FHF).
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Endotoxin Content:<1 EU/μg by LAL method
- Reconstitution Instructions:Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 µg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.
- Protein Synonyms:TNFRSF1A
- Protein/Peptide Name:TNFR1
- Purity:>92%
- Molecular Weight:22 kDa
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.