>9% pure active human cell expressed human recombinant HAVCR1/KIM1/TIM1
Hepatitis A virus cellular receptor 1 also known as HAVCR1, HAVCR, KIM1, TIM, TIM1, TIMD1, is widely expressed with highest levels in kidney and testis. The protein encoded by HAVCR1 gene is a membrane receptor for both human hepatitis A virus (HHAV) and TIMD4. The encoded protein may be involved in the moderation of asthma and allergic diseases. The reference genome represents an allele that retains a MTTVP amino acid segment that confers protection against atopy in HHAV seropositive individuals. Three transcript variants encoding the same protein have been found for this gene. HAVCR1 may play a role in T-helper cell development, the regulation of asthma and allergic diseases and in kidney injury and repair. In case of human hepatitis A virus (HHAV) infection, functions as a cell-surface receptor for the virus.
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Endotoxin Content:<1 EU/μg by LAL method
- Reconstitution Instructions:Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 µg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.
- Protein Synonyms:TIM1|TIM-1|KIM-1|HAVCR|Hepatitis A virus cellular receptor 1|HAVCR-1|KIM1
- Protein/Peptide Name:HAVCR1
- Purity:>90%
- Molecular Weight:31.3 kDa
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.