Order Entry
United States
ContactUsLinkComponent
Human Recombinant GPT2 (from E. coli)
Human Recombinant GPT2 (from E. coli)
Catalog # 10835-458
Supplier:  Avantor
CAS Number:  
undefined
Human Recombinant GPT2 (from E. coli)
Catalog # 10835-458
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 μg
  • Environmentally Preferable:
  • Protein Synonyms:
    Al
  • Protein/Peptide Name:
    GPT2
  • Purity:
    >90%
  • Molecular Weight:
    60.3 kDa
  • Formulation:
    0.5 mg/ml solution 20 mM Tris-HCl buffer (pH 7.5), 30% glycerol, 0.2 M NaCl and 2 mM DTT.
  • Cat. No.:
    10835-458
  • Supplier no.:
    7806-50

Specifications

About this item

≥9% Pure active Recombinant Human GPT2/ALT2

Alanine aminotransferase 2, also known as GPT2, catalyzes the reversible transamination between alanine and 2-oxoglutarate to form pyruvate and glutamate. Subsequently, they play a key role in the intermediary metabolism of glucose and amino acids. GPT2 expression is high in muscle, fat and kidney. Recombinant human GPT2 protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.