ContactUsLinkComponent
Staticide® Turbo Blast Precision Duster, ACL
Staticide® Turbo Blast Precision Duster, ACL
Catalog # 75780-007
Supplier:  ACL Staticide
undefined
Staticide® Turbo Blast Precision Duster, ACL
Catalog # 75780-007
Supplier:  ACL Staticide
Supplier Number:  8640
Specifications
  • Volume:
    296 g (11 oz.)
  • Description:
    Staticide® Turbo Blast Precision Duster
  • Cat. No.:
    75780-007
Specifications
About this item
Staticide® Turbo Blast Precision Duster is an economical, high-precision duster that offers maximum blasting power for electronic cleaning applications.
Filtered to 0.2 microns, this duster provides exceptional purity, and rids surfaces of particulate contamination without leaving residuals. Engineered to remove dirt, dust, oxide particles, and other airborne particles instantly. It is also excellent in accelerating the drying time of solvents. Includes straw for even greater pinpoint blasting.