VascuLife® VEGF-Mv is optimized for the culture of human microvascular endothelial cells, and supports the rapid proliferation of these cells in a 5% serum environment.
- Optimized for the culture of human microvascular endothelial cells
- Supports rapid cell proliferation
- Contains no antimicrobials or phenol red
The VascuLife® VEGF-Mv LifeFactors® Kit includes: rh FGF basic 0.5 mL, Ascorbic Acid 0.5 mL, Hydrocortisone Hemisuccinate 0.5 mL, L-Glutamine 25 mL, rh IGF-1 0.5 mL, rh EGF 0.5 mL, rh VEGF 0.5 mL, Heparin Sulfare 0.5 mL, FBS 25 mL, Gentamicin Amphotericin B 0.5 mL.