About this item
VascuLife® EnGS-Mv is optimized for the culture of human microvascular endothelial cells, and supports the rapid proliferation of these cells in a 5% serum environment.
- Optimized for the rapid proliferation of human microvascular endothelial cells
- 5% serum environment
- Maintains excellent cell morphology
The VascuLife® EnGS-Mv Medium Complete Kit includes: VascuLife® Basal Medium (475 mL) and VascuLife® EnGS-Mv LifeFactors® Kit.VascuLife® contains no antimicrobials and no phenol red. VascuLife® EnGS-Mv (containing Endothelial Cell Growth Supplement) is optimized for the rapid proliferation of human microvascular endothelial cells without VEGF, phenol red, or antimicrobials, at rates that meet or exceed other commercially available serum-containing media while maintaining excellent cell morphology.
The VascuLife® EnGS-Mv LifeFactors® Kit includes: EnGS 1.0 mL, rh EGF 0.5 mL, Ascorbic Acid 0.5 mL, L-Glutamine 25 mL, Hydrocortisone Hemisuccinate 0.5 mL, Heparin Sulfate 0.5 mL, FBS 10 mL, Gentamicin and Amphotericin B 0.5 mL.