ContactUsLinkComponent
FibroLife® Xeno-Free Frozen Medium, Lifeline Cell Technology
FibroLife® Xeno-Free Frozen Medium, Lifeline Cell Technology
  102970-882
 :  
undefined
FibroLife® Xeno-Free Frozen Medium, Lifeline Cell Technology
  102970-882
 :  LM-0013
 :  
  • Description:
    FibroLife® Xeno-Free Complete Frozen Medium
  • Size:
    500 mL
  • Cat. No.:
    102970-882
  • Supplier no.:
    LM-0013
FibroLife® Xeno-Free Medium is optimized for culture of fibroblasts without the use of non-human animal-derived components. FibroLife® Xeno-Free supports the growth of these cells in a 2% human serum environment at rates equal to or greater than comparable media supplemented with 2-10% FBS. FibroLife® contains no phenol red or antimicrobials to affect cell performance.
  • Extended shelf life of up to two years
  • Optimized for culture of fibroblasts
  • Contains no phenol red or antimicrobials to affect cell performance
FibroLife® Xeno-Free Medium is optimized for culture of fibroblasts without the use of non-human animal-derived components
FibroLife® Xeno-Free is offered as a fully-supplemented Complete Frozen Medium with extended shelf-life of up to two years.
FibroLife® Xeno-Free is also offered as a Medium Complete Kit that contains: FibroLife® Xeno-Free Complete Frozen Medium 500 mL, Phosphate Buffered Saline (Xeno-Free) 500 mL, Trypsin/EDTA Xeno-Free 100 mL, Trypsin Neutralizing Solution Xeno-Free 100 mL, FrostaLife™ Xeno-Free Cryopreservation Solution 100 mL, Xeno-Free Antimicrobial Supplement: Penicillin/Streptomycin 1.0 mL.
FibroLife Xeno-Free is available as a fully-supplemented Frozen Medium by itself, or with associated Xeno-Free and other products in a Complete Kit.