- Description:AimStep® H. Pylori Test Kit
- Cat. No.:10024-408
- Contents:Test Device, Developer Solution, Positive & Negative Controls, Transfer Pipettes, and Procedure Card
- Supplier no.:33020
- No. of tests:20
Cassette method for the qualitative detection of anti-Helicobacter pylori antibody in human whole blood, serum, or plasma
This test is intended to aid in the diagnosis of H. pilori infection in adult patients with symptoms of gastrointestinal disorders.