ContactUsLinkComponent
AimStep® Mono Test Kit, Germaine® Laboratories
AimStep® Mono Test Kit, Germaine® Laboratories
Catalog # 10024-406
CAS Number:  
undefined
AimStep® Mono Test Kit, Germaine® Laboratories
Catalog # 10024-406
Supplier Number:  77025
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    AimStep® Mono Test Kit
  • Cat. No.:
    10024-406
  • Contents:
    Test Device, Developer Solution, Positive & Negative Controls, Transfer Pipettes, and Procedure Card
  • Supplier no.:
    77025
  • No. of tests:
    25
Specifications
About this item
Immunoassay for the qualitative detection of infectious Mononucleosis Heterophile Antibodies in whole blood, serum, or plasma
Cassette method.
Certifications: CLIA waived for whole blood. CLIA moderately complex for serum and plasma.