hCG Controls, Germaine® Laboratories
Supplier: Germaine Laboratories
Certificates
About this item
Liquid controls for quality testing of pregnancy tests
The hCG Urine Controls are bi-level controls for use with urine pregnancy tests. The hCG Serum Controls are tri-level controls for use with serum pregnancy tests.
Frequently Bought Together