- Material:Steel, nickel plated, #4
- Cat. No.:10170-208
- For:Blade no.: 20 - 25
- Supplier no.:53410
Securely hold blades with the Shandon™ nickel-plated scalpel blade handle, constructed of rustless, nickel-plated steel #4.
- Properly hardened, tempered and passivated
- Electronically polished and cleaned for a high-quality finish
- Defect-free in material and workmanship
Quality patterns, dimensions and finish held to exacting standards, producing the finest instruments available.