Order Entry
United States
ContactUsLinkComponent
Epredia™ Cassette Sponge, Epredia
Epredia™ Cassette Sponge, Epredia
Catalog # 89404-854
Supplier:  Epredia
undefined
Epredia™ Cassette Sponge, Epredia
Catalog # 89404-854
Supplier:  Epredia
Supplier Number:  8453

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Color:
    Blue
  • Description:
    Rectangular sponge
  • Dimensions:
    25×31 mm (1×1.2")
  • Cat. No.:
    89404-854
  • Supplier no.:
    8453

Specifications

About this item

Protect small biopsies from overprocessing with the Epredia™ Sponge.

  • Blue, open-cell foam sponge holds specimen in cassette