ContactUsLinkComponent
Castroviejo Lachrymal Dilator, OR Grade, Sklar®
Castroviejo Lachrymal Dilator, OR Grade, Sklar®
  10197-422
 :  Sklar
undefined
Castroviejo Lachrymal Dilator, OR Grade, Sklar®
  10197-422
 :  Sklar
 :  65-6960
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Castroviejo Lacrimal Dilator, Delicate
  • Length:
    4¾"
  • Material:
    Stainless steel
  • Tip configuration:
    Taper and sharp
  • Curvature:
    Straight
  • Cat. No.:
    10197-422
  • Supplier no.:
    65-6960
This is a double-ended instrument most commonly used in ophthalmologic, lacrimal system disorder procedures, and oculofacial plastic surgery.
  • Straight and round
  • Two tapered tips
The lachrymal dilator may be used to open the tear duct to increase tear flow, access the lachrymal system for obstruction removal, and repair damage to or disease of the lachrymal system.