About this item
p57 Kip2 (or CDKN1C) is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. The gene encoding human p57 Kip2 is located on chromosome 11p15.5, a region implicated in both sporadic cancers, Wilm's tumor, and Beckwith-Wiedemann syndrome (BWS), a cancer syndrome, making it a tumor suppressor candidate. BWS is characterized by numerous growth abnormalities and an increased risk of childhood tumors. Several types of childhood tumors including Wilms' tumor, adrenocortical carcinoma and rhabdomyosarcoma display a specific loss of maternal 11p15 alleles, suggesting that genomic imprinting plays an important part. This region also contains two other imprinted genes, insulin-like growth factor II (IGF-II) and H19, both of which seem to be implicated in adrenal neoplasms. Positive Control: LS174T cells, Colon CA or placenta; Cellular Localization: Nuclear; Recommended for: Immunohistochemistry (Formalin/paraffin), Western Blotting (Not suitable), Immunoprecipitation (Native verified)
Type: Primary
Antigen: P57KIP2
Clonality: Monoclonal
Clone: 57P06
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 100157-072
- 10170-242
- Antigen Symbol:
- P57KIP2
- P57KIP2
- Antigen Name:
- Cyclin-dependent kinase inhibitor 1C
- Cyclin-dependent kinase inhibitor 1C
- Antigen Synonyms:
- Cyclin-dependent kinase inhibitor p57|p57Kip2|Cyclin-dependent kinase inhibitor 1C
- Cyclin-dependent kinase inhibitor p57|p57Kip2|Cyclin-dependent kinase inhibitor 1C
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 57P06
- 57P06
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- Recombinant human p57 Kip2 protein
- Molecular Weight:
- 57 kDa