- Description:Vapor Pad
- Dimensions:11 x 14"
- Environmentally Preferable:
- Cat. No.:10809-806
- Supplier no.:VP01114
Formalin Neutralizer Pads encapsulate and neutralize harmful vapors to greatly reduce exposure to Formaldehyde.
- Offers contrast for all biopsies
- Cut resistant
- Durable
- Disposable
- Neutralizes harmful vapors
Formalin Neutralizer Pads that encapsulate and neutralize harmful vapors to greatly reduce exposure to Formaldehyde. Light blue contained work surface offers contrast for all biopsies. Cut resistant, durable and disposable.