Order Entry
United States
Orders LinkContactUsLinkComponent
 

Stacking Container, MFG Tray

Stacking Container, MFG Tray
Certificates
Stacking Container, MFG Tray

11 Options Available Below

Product Details & Documents

MFG Tray's durable Stacking containers are resistant to cutting oils, grease, and solvents

Easy to clean with hot water or steam using standard detergents. Ideal for forgings, machined parts, stampings, ceramics, rubbers, and other heavy or hot parts emerging from molds or processing machines. Perfect for work-in-process part handling. MFG Tray is a leading manufacturer of high-strength, glass-reinforced composite containers, trays, boxes, flats, and totes for material handling in the metalworking, plastics, and electronics assembly industries, as well as in food service, confectionery, and pharmaceutical processing. MFG Tray products combine outstanding performance features with long service life to ensure exceptional value. The material matters in material handling.

Specifications for Product Family

Specifications

  • Catalog No:
  • 89022-902
  • 89022-888
  • 89022-900
  • 89022-890
  • 89022-892
  • 89022-898
  • 89022-896
  • 89022-904
  • 89022-414
  • 89177-122
  • 89022-910
  • pH Range:
  • 3.0 to 10.0
  • 3.0 to 10.0
  • 3.0 to 10.0
  • 3.0 to 10.0
  • 3.0 to 10.0
  • 3.0 to 10.0
  • 3.0 to 10.0
  • 3.0 to 10.0
  • 3.0 to 10.0
  • 3.0 to 10.0
  • 3.0 to 10.0
  • Temperature Range:
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • –51 to 121°C (–60 to 250°F), up to 300°F intermittently

Specifications

Product Family Options

Product Information

  • Dimensions
    Color
    Availability
    Price
  • 89022-902
    Stacking Container
    Stacking Container
    14.4 x 14.4 x 3.5"
    White
    Log in for availability
    Log in for availability
    Each
    Each - $82.09
    $82.09
    Each
       $82.09
     

    Supplier Number: 825008-5269
    Specifications:
    More
    Specifications
    Cat. No.
    89022-902
    pH Range
    3.0 to 10.0
    Temperature Range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • 89022-888
    Stacking Container
    Stacking Container
    16.5 x 1 x 4.6"
    White
    Log in for availability
    Log in for availability
    Each
    Each - $65.86
    $65.86
    Each
       $65.86
     

    Supplier Number: 802008-5269
    Specifications:
    More
    Specifications
    Cat. No.
    89022-888
    pH Range
    3.0 to 10.0
    Temperature Range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • 89022-900
    Stacking Container
    Stacking Container
    17.5 x 17.5 x 4.5"
    White
    Log in for availability
    Log in for availability
    Each
    Each - $91.93
    $91.93
    Each
       $91.93
     

    Supplier Number: 817008-5269
    Specifications:
    More
    Specifications
    Cat. No.
    89022-900
    pH Range
    3.0 to 10.0
    Temperature Range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • 89022-890
    Stacking Container
    Stacking Container
    18.75 x 11.8 x 1.75"
    White
    Log in for availability
    Log in for availability
    Each
    Each - $57.37
    $57.37
    Each
       $57.37
     

    Supplier Number: 804008-5269
    Specifications:
    More
    Specifications
    Cat. No.
    89022-890
    pH Range
    3.0 to 10.0
    Temperature Range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • 89022-892
    Stacking Container
    Stacking Container
    22 x 12 x 1.25"
    White
    Log in for availability
    Log in for availability
    Each
    Each - $54.46
    $54.46
    Each
       $54.46
     

    Supplier Number: 808108-5269
    Specifications:
    More
    Specifications
    Cat. No.
    89022-892
    pH Range
    3.0 to 10.0
    Temperature Range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • 89022-898
    Stacking Container
    Stacking Container
    23.375 x 12 x 4.375"
    White
    Log in for availability
    Log in for availability
    Each
    Each - $85.18
    $85.18
    Each
       $85.18
     

    Supplier Number: 808408-5269
    Specifications:
    More
    Specifications
    Cat. No.
    89022-898
    pH Range
    3.0 to 10.0
    Temperature Range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • 89022-896
    Stacking Container
    Stacking Container
    23.375 x 12 x 6"
    White
    Log in for availability
    Log in for availability
    Each
    Each - $102.33
    $102.33
    Each
       $102.33
     

    Supplier Number: 808308-5269
    Specifications:
    More
    Specifications
    Cat. No.
    89022-896
    pH Range
    3.0 to 10.0
    Temperature Range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • 89022-904
    Stacking Container
    Stacking Container
    25.75 x 17.75 x 3"
    White
    Log in for availability
    Log in for availability
    Each
    Each - $93.55
    $93.55
    Each
       $93.55
     

    Supplier Number: 870008-5269
    Specifications:
    More
    Specifications
    Cat. No.
    89022-904
    pH Range
    3.0 to 10.0
    Temperature Range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • 89022-414
    Stacking Container
    Stacking Container
    White
    Log in for availability
    Log in for availability
    Each
    Each - $100.20
    $100.20
    Each
       $100.20
     

    Supplier Number: 875008-5269
    Specifications:
    More
    Specifications
    Cat. No.
    89022-414
    pH Range
    3.0 to 10.0
    Temperature Range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • 89177-122
    Lid for 808208, 808308, and 808408 series Stacking Containers
    Lid for 808208, 808308, and 808408 series Stacking Containers
    White
    Log in for availability
    Log in for availability
    Each
    Each - $52.17
    $52.17
    Each
       $52.17

    Supplier Number: 8082185269
    Specifications:
    More
    Specifications
    Cat. No.
    89177-122
    pH Range
    3.0 to 10.0
    Temperature Range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently
  • 89022-910
    Lid for 870008, 875008, and 880008 series Stacking Containers
    Lid for 870008, 875008, and 880008 series Stacking Containers
    White
    Log in for availability
    Log in for availability
    Each
    Each - $87.50
    $87.50
    Each
       $87.50
     

    Supplier Number: 887008-5269
    Specifications:
    More
    Specifications
    Cat. No.
    89022-910
    pH Range
    3.0 to 10.0
    Temperature Range
    –51 to 121°C (–60 to 250°F), up to 300°F intermittently