Rinsing Solution Fluid D Clean Rooms, bioMerieux
Catalog # 97067-036
Supplier: bioMerieux
CAS Number:
Specifications
- Description:Rinsing Solution Fluid D Clean Rooms, Bottles
- Cat. No.:97067-036
- Supplier no.:42644
Specifications
About this item
Rinsing Solution Fluid D Clean Rooms is a sterile diluent for rinsing membranes during sterility testing.
- Double wrapped
- Ethylene Oxide decontaminated
- Membrane filtration
For sterility testing. Double wrapped and Ethylene Oxide decontaminated.