Order Entry
United States
ContactUsLinkComponent
Buffered Peptoned Solution, pH 7.0, bioMerieux
Buffered Peptoned Solution, pH 7.0, bioMerieux
Catalog # 95060-774
Supplier:  bioMerieux
CAS Number:  
Buffered Peptoned Solution, pH 7.0, bioMerieux
Catalog # 95060-774
Supplier:  bioMerieux
Supplier Number:  42609
CAS Number:  

Specifications

  • Description:
    Buffered Peptone Solution
  • Size:
    90 mL
  • Cat. No.:
    95060-774
  • pH:
    7
  • Supplier no.:
    42609

Specifications

About this item

Buffered Peptoned Solution detects and enumerates microorganisms.

  • Diluent
  • Detects and enumerates microorganisms

Media for general microbiology testing.