To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:HDL Cholesterol E
- Size:1 x 20 mL, 2 x 150 mL (kit)
- Storage Temperature:2-10° C
- Cat. No.:10752-436
- Supplier no.:431-52501
Specifications
About this item
The HDL Cholesterol E kit determines HDL Cholesterol in serum.
- Utilizes a manual precipitation method
- Determines HDL Cholesterol in serum
- 90 tests
The HDL Cholesterol E kit contains reagents utilizing a manual precipitation method for the determination of HDL Cholesterol in serum. Contains 90 tests.