ContactUsLinkComponent
Human Vas Deferens Fibroblasts, Lifeline Cell Technology
Human Vas Deferens Fibroblasts, Lifeline Cell Technology
Catalog # 102970-934
CAS Number:  
undefined
Human Vas Deferens Fibroblasts, Lifeline Cell Technology
Catalog # 102970-934
Supplier Number:  FC-0052
CAS Number:  
Specifications
  • Cell Format:
    Cryopreserved
  • Description:
    Human Vas Deferens Fibroblasts
  • Size:
    500,000 cells per vial
  • Cat. No.:
    102970-934
  • Supplier no.:
    FC-0052
Specifications
About this item
Lifeline® Normal Human Vas Deferens Fibroblasts need no exposure to antimicrobials or phenol red when cultured in Lifeline® FibroLife® media.
  • Population growth at various densities
  • Exposure to antimicrobials or phenol red unnecessary
  • Negative for mycoplasma
  • Negative for bacterial and fungal growth
Lifeline® Normal Human Vas Deferens Fibroblasts need not be exposed to antimicrobials or phenol red when cultured in Lifeline® FibroLife® media.
Vas Deferens Fibroblasts are tested for: a) Proliferation: Population growth at various densities, b) Cell Viability: 70% or greater when thawed from cryopreservation, c) Morphology: Normal cell appearance for 15 population doublings, d) Sterility: Negative for mycoplasma and Negative for bacterial and fungal growth, and e) Virus: Negative for HIV-1, HIV-2, HBV, and HCV by PCR.