ContactUsLinkComponent
Human Lung Fibroblasts, Lifeline Cell Technology
Human Lung Fibroblasts, Lifeline Cell Technology
Catalog # 102970-702
CAS Number:  
undefined
Human Lung Fibroblasts, Lifeline Cell Technology
Catalog # 102970-702
Supplier Number:  FC-0049
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Cell Format:
    Cryopreserved
  • Description:
    Human Lung Fibroblasts
  • Size:
    500,000 cells per vial
  • Cat. No.:
    102970-702
  • Supplier no.:
    FC-0049
Specifications
About this item
Lifeline® Normal Human Lung Fibroblasts are quality tested and demonstrate optimal growth.
  • Quality tested
  • Optimal growth
  • Exposure to antimicrobials or phenol red unnecessary
Lifeline® Normal Human Lung Fibroblasts are quality tested in FibroLife® S2 or FibroLife® Xeno-Free Medium and demonstrate optimal growth at rates equal to or greater than classical serum-supplemented media
Lifeline® Human Lung Fibroblasts need not be exposed to antimicrobials or phenol red when cultured in FibroLife® Medium.
Fibroblasts are tested for: