ContactUsLinkComponent
Extract-P LifeFactor, Bovine, Lifeline Cell Technology
Extract-P LifeFactor, Bovine, Lifeline Cell Technology
Catalog # 102970-850
CAS Number:  
Extract-P LifeFactor, Bovine, Lifeline Cell Technology
Catalog # 102970-850
Supplier Number:  LS-1037
CAS Number:  
Specifications
  • Description:
    Extract-P LifeFactor
  • Size:
    2 mL
  • Cat. No.:
    102970-850
  • Supplier no.:
    LS-1037
Specifications
About this item
Extract P may be used with basal medium, and human cells and keratinocytes.
  • Used with basal medium
  • Used with human reproductive cells
  • Used with human corneal epithelial cells
  • Used with human airway epithelial cells
  • Used with human mammary epithelial cells
  • Used with human bladder apex epithelial cells
Extract P may be used with these Lifeline® culture media and human cell types:
ProstaLife™ basal medium and human reproductive epithelial cells, DermaLife basal medium or DermaLife calcium-free basal medium and human keratinocytes, OcuLife™ basal medium and human corneal epithelial cells, UroLife™ A basal medium and human bladder apex epithelial cells, BronchiaLife™ basal medium and human airway epithelial cells, MammaryLife™ basal medium and human mammary epithelial cells.