ContactUsLinkComponent
L-Glutamine LifeFactor, Lifeline Cell Technology
L-Glutamine LifeFactor, Lifeline Cell Technology
Catalog # 102970-732
CAS Number:  
L-Glutamine LifeFactor, Lifeline Cell Technology
Catalog # 102970-732
Supplier Number:  LS-1013
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    L-Glutamine LifeFactor
  • Size:
    25 mL
  • Cat. No.:
    102970-732
  • Supplier no.:
    LS-1013
Specifications
About this item
L-Glutamine LifeFactor is used with basal medium and human cells.
  • Used with human endothelial cells
  • Used with human smooth muscle cells
  • Used with basal medium
L-Glutamine LifeFactor -- 25 mL may be used with: Lifeline® VascuLife® basal medium and human smooth muscle cells, or Lifeline® VascuLife® basal medium and human endothelial cells.