ContactUsLinkComponent
Human Lung Smooth Muscle Cells, Lifeline Cell Technology
Human Lung Smooth Muscle Cells, Lifeline Cell Technology
Catalog # 102970-698
CAS Number:  
undefined
Human Lung Smooth Muscle Cells, Lifeline Cell Technology
Catalog # 102970-698
Supplier Number:  FC-0046
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Cell Format:
    Cryopreserved
  • Description:
    Human Lung Smooth Muscle Cells
  • Size:
    500,000 cells per vial
  • Cat. No.:
    102970-698
  • Supplier no.:
    FC-0046
Specifications
About this item
Lifeline® Normal Human Lung Smooth Muscle Cells are quality tested and demonstrate optimal low serum growth.
  • Quality tested in VascuLife® SMC Medium
  • Demonstrate optimal low serum growth
  • Can be grown without antimicrobials or phenol red
Lifeline® Normal Human Lung Smooth Muscle Cells are quality tested in VascuLife® SMC Medium and demonstrate optimal low serum growth over a period of at least 15 population doublings at rates equal to or greater than other serum-supplemented media.
Lung Smooth Muscle Cells can be grown without phenol red or antimicrobials when cultured in VascuLife® SMC Medium.