Specifications
- Description:Hydrocortisone Hemisuccinate LifeFactor
- Size:1 mg/mL
- Cat. No.:102970-982
- Supplier no.:LS-1007
Specifications
About this item
Hydrocortisone Hemisuccinate is used with fibroblasts, basal medium, and endothelial cells.
- Used with human fibroblasts
- Used with human endothelial cells
- Used with basal medium
Hydrocortisone Hemisuccinate LifeFactor may be used with: FibroLife®basal medium and human fibroblasts, VascuLife® basal medium and human endothelial cells.